Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fipr-5
Entrez Gene ID
3565026
Gene Name
FIP (Fungus-Induced Protein) Related
Gene Aliases
CELE_C12D8.18
Chromosome Location
Chr.V: 10220981 - 10221417 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028483.2 NP_001023654.1 1 - 2 134 108

View Full Product Details