Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ttll-9
Entrez Gene ID
3564837
Gene Name
Probable tubulin polyglutamylase ttll-9
Gene Aliases
CELE_F25C8.5
Chromosome Location
Chr.V: 20910467 - 20914387 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028670.2 NP_001023841.1 2 - 3 222 76

View Full Product Details