Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-11
Entrez Gene ID
3565837
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_T09F5.16
Chromosome Location
Chr.V: 15163082 - 15164518 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028955.1 NP_001024126.1 3 - 4 717 126

View Full Product Details