Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-36
Entrez Gene ID
3565454
Gene Name
C-type LECtin
Gene Aliases
CELE_T20B3.16
Chromosome Location
Chr.V: 16846811 - 16848598 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028979.4 NP_001024150.2 5 - 6 708 59

View Full Product Details