Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tsp-21
Entrez Gene ID
259720
Gene Name
Tetraspanin-21
Gene Aliases
CELE_C17G1.8
Chromosome Location
Chr.X: 9928285 - 9937133 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383631.1 7 - 8 751 68

View Full Product Details