Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-19
Entrez Gene ID
180397
Gene Name
Cuticle collagen 19
Gene Aliases
CELE_ZK1193.1
Chromosome Location
Chr.X: 421707 - 423689 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038488.5 NP_001033577.1 1 - 2 150 77

View Full Product Details