Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srz-6
Entrez Gene ID
188671
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_T21B4.1
Chromosome Location
Chr.II: 12492236 - 12493627 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001368611.2 NP_001355366.1 2 - 3 931 67

View Full Product Details