Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
adpr-1
Entrez Gene ID
3565128
Gene Name
protein-coding
Gene Aliases
CELE_Y73E7A.9
Chromosome Location
Chr.I: 1610390 - 1621335 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026658.2 NP_001021829.1 7 - 8 1363 152
NM_001306479.3 NP_001293408.1 4 - 5 739 152
NM_001306480.3 NP_001293409.1 3 - 4 586 152
NM_001306481.3 NP_001293410.1 2 - 3 238 152

View Full Product Details