Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-28
Entrez Gene ID
3565225
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_D2062.12
Chromosome Location
Chr.II: 2625092 - 2627109 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026882.3 NP_001022053.1 5 - 6 760 119

View Full Product Details