Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-42
Entrez Gene ID
173503
Gene Name
protein-coding
Gene Aliases
CELE_F47F6.1
Chromosome Location
Chr.II: 1233235 - 1252359 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027005.4 NP_001022176.1 4 - 5 1119 114
NM_001027006.6 NP_001022177.1 8 - 9 1552 114
NM_001322704.3 NP_001309559.1 9 - 10 1703 114
NM_001322705.3 NP_001309560.1 4 - 5 749 114

View Full Product Details