Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oig-4
Entrez Gene ID
259448
Gene Name
Immunoglobulin domain-containing protein oig-4
Gene Aliases
CELE_R07G3.9
Chromosome Location
Chr.II: 7613541 - 7614435 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027107.5 NP_001022278.1 3 - 4 457 110

View Full Product Details