Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-23
Entrez Gene ID
3565992
Gene Name
Zinc metalloproteinase nas-23
Gene Aliases
CELE_R10H1.5
Chromosome Location
Chr.II: 7491267 - 7493699 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027110.3 NP_001022281.1 2 - 3 218 79

View Full Product Details