Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-34
Entrez Gene ID
3565058
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_W05H5.8
Chromosome Location
Chr.II: 12450149 - 12451407 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027224.1 NP_001022395.1 1 - 2 465 105

View Full Product Details