Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
glb-29
Entrez Gene ID
189454
Gene Name
protein-coding
Gene Aliases
CELE_Y17G7B.6
Chromosome Location
Chr.II: 12009072 - 12023832 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267402.3 NP_001254331.1 2 - 3 210 108
NM_001267403.3 NP_001254332.1 2 - 3 237 108

View Full Product Details