Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-7
Entrez Gene ID
175213
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_T19C3.9
Chromosome Location
Chr.III: 606004 - 606998 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027599.7 NP_001022770.1 2 - 3 304 91

View Full Product Details