Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srw-9
Entrez Gene ID
3565297
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_ZC482.8
Chromosome Location
Chr.III: 12757667 - 12759619 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027806.1 NP_001022977.1 1 - 2 592 82

View Full Product Details