Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srz-71
Entrez Gene ID
3564811
Gene Name
Serpentine Receptor, class Z
Gene Aliases
CELE_C35D6.10
Chromosome Location
Chr.IV: 16332885 - 16334461 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027877.2 NP_001023048.2 1 - 2 154 153

View Full Product Details