Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-15
Entrez Gene ID
3565991
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C39H7.8
Chromosome Location
Chr.IV: 5622063 - 5623818 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027883.2 NP_001023054.2 3 - 4 466 108

View Full Product Details