Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-8
Entrez Gene ID
3565159
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_C43F9.10
Chromosome Location
Chr.IV: 10584537 - 10587548 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027890.2 NP_001023061.1 5 - 6 853 69

View Full Product Details