Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hrg-6
Entrez Gene ID
3565935
Gene Name
protein-coding
Gene Aliases
CELE_F36H1.10
Chromosome Location
Chr.IV: 11032588 - 11033599 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268599.2 NP_001255528.1 1 - 2 209 60

View Full Product Details