Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-8
Entrez Gene ID
182605
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C14C6.9
Chromosome Location
Chr.V: 554374 - 556155 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070843.3 NP_503244.2 3 - 4 449 148

View Full Product Details