Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srbc-2
Entrez Gene ID
182214
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_C04E12.9
Chromosome Location
Chr.V: 3374362 - 3377073 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071539.5 NP_503940.2 2 - 3 342 118

View Full Product Details