Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nspc-4
Entrez Gene ID
3565092
Gene Name
Nematode Specific Peptide family, group C
Gene Aliases
CELE_C03G5.11
Chromosome Location
Chr.X: 8537956 - 8538374 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029177.2 NP_001024348.1 2 - 2 156 125

View Full Product Details