Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
puf-6
Entrez Gene ID
174947
Gene Name
Pumilio domain-containing protein 6
Gene Aliases
CELE_F18A11.1
Chromosome Location
Chr.II: 13039790 - 13043445 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064372.7 NP_496773.1 3 - 4 1251 152

View Full Product Details