Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dct-7
Entrez Gene ID
184536
Gene Name
DAF-16/FOXO Controlled, germline Tumor affecting
Gene Aliases
CELE_F15E6.8
Chromosome Location
Chr.IV: 4290189 - 4290703 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068133.2 NP_500534.1 1 - 2 253 119

View Full Product Details