Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbxa-85
Entrez Gene ID
4363110
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y61B8A.4
Chromosome Location
Chr.V: 17259168 - 17260561 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047735.2 NP_001041200.1 2 - 3 738 145

View Full Product Details