Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ttr-38
Entrez Gene ID
4363115
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_F35G8.2
Chromosome Location
Chr.X: 9312515 - 9314497 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047783.3 NP_001041248.1 2 - 3 287 93

View Full Product Details