Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gsto-2
Entrez Gene ID
353420
Gene Name
Probable glutathione transferase omega-2
Gene Aliases
CELE_C02D5.3
Chromosome Location
Chr.III: 8554772 - 8555744 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181976.5 NP_871705.3 2 - 3 534 151

View Full Product Details