Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
set-29
Entrez Gene ID
171705
Gene Name
SET domain-containing protein
Gene Aliases
CELE_Y92H12BR.6
Chromosome Location
Chr.I: 1405935 - 1413100 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058448.6 NP_490849.4 2 - 3 625 153

View Full Product Details