Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
QKI
Entrez Gene ID
606754
Gene Name
QKI, KH domain containing RNA binding
Gene Aliases
CQKI, QKI-5
Chromosome Location
Chr.1: 51800294 - 51956891 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003021.2 NP_001003021.1 - 648 61
GenBank mRNA AB047312.1 - - 652 61

View Full Product Details