Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGA
Entrez Gene ID
403711
Gene Name
pepsinogen 5, group I (pepsinogen A)
Gene Aliases
PGA5, PGNA
Chromosome Location
Chr.18: 55131441 - 55140119 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003117.1 NP_001003117.1 3 - 4 371 60
GenBank mRNA AB047246.1 - 3 - 4 371 60

View Full Product Details