Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CELA1
Entrez Gene ID
403515
Gene Name
chymotrypsin like elastase 1
Gene Aliases
ELA1, ELS1
Chromosome Location
Chr.27: 3542327 - 3560777 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003007.2 NP_001003007.2 5 - 6 515 76
GenBank mRNA AB100595.1 - 4 - 5 469 76
DN357055.1 - 5 - 6 536 76
DN357380.1 - 5 - 6 514 76
DN359987.1 - 5 - 6 485 76

View Full Product Details