Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSP70
Entrez Gene ID
403612
Gene Name
heat shock protein 70
Gene Aliases
HSPA1
Chromosome Location
Chr.12: 1285282 - 1287670 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003067.2 NP_001003067.2 1 - 1 2083 69
GenBank mRNA AB075027.1 - 1 - 1 1998 69
AB114672.1 - 1 - 1 1982 69
AB114673.1 - 1 - 1 1969 69
AB114674.1 - 1 - 1 1976 69
AB114675.1 - 1 - 1 1975 69

View Full Product Details