Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDC
Entrez Gene ID
403624
Gene Name
phosducin
Gene Aliases
-
Chromosome Location
Chr.7: 19498785 - 19514225 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003076.1 NP_001003076.1 1 - 2 60 113
GenBank mRNA AF046874.1 - 1 - 2 60 113
Y17697.1 - - 49 113

View Full Product Details