Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD28
Entrez Gene ID
403646
Gene Name
CD28 molecule
Gene Aliases
-
Chromosome Location
Chr.37: 12503849 - 12536362 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003087.2 NP_001003087.2 1 - 2 135 62
GenBank mRNA AF154842.1 - 1 - 2 135 62
AF259962.1 - 1 - 2 174 62

View Full Product Details