Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHBG
Entrez Gene ID
403538
Gene Name
Rh family B glycoprotein
Gene Aliases
-
Chromosome Location
Chr.7: 41532266 - 41548523 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003017.2 NP_001003017.2 7 - 8 1171 58
XM_005622221.3 XP_005622278.1 8 - 9 1364 58
XM_022420725.1 XP_022276433.1 8 - 9 1367 58
GenBank mRNA AY622224.1 - 7 - 8 1171 58

View Full Product Details