Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ELSPBP1
Entrez Gene ID
399530
Gene Name
epididymal sperm binding protein 1
Gene Aliases
E12, E12A
Chromosome Location
Chr.1: 108159609 - 108187224 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002931.1 NP_001002931.1 1 - 2 60 80
XM_005615714.3 XP_005615771.1 1 - 2 155 80
XM_005615715.3 XP_005615772.1 1 - 2 157 80
XM_022404230.1 XP_022259938.1 1 - 2 163 80
XM_022404725.1 XP_022260433.1 1 - 2 161 80
XM_022427808.1 XP_022283516.1 1 - 2 159 80
GenBank mRNA AJ278477.1 - 1 - 2 60 80

View Full Product Details