Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRAS
Entrez Gene ID
403872
Gene Name
NRAS proto-oncogene, GTPase
Gene Aliases
N-RAS, rasp21
Chromosome Location
Chr.17: 52411733 - 52418788 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287065.1 NP_001273994.1 2 - 3 200 82
GenBank mRNA U62093.1 - 1 - 2 86 82

View Full Product Details