Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTSD
Entrez Gene ID
483662
Gene Name
cathepsin D
Gene Aliases
-
Chromosome Location
Chr.18: 46010440 - 46019457 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025621.1 NP_001020792.1 7 - 8 1037 60
GenBank mRNA AM048627.1 - 7 - 8 1037 60

View Full Product Details