Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SULT1A1
Entrez Gene ID
403892
Gene Name
sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1
Gene Aliases
PST, ST1A1, cSULT1A1
Chromosome Location
Chr.6: 18176156 - 18179492 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003223.1 NP_001003223.1 5 - 6 576 73
XM_014114387.1 XP_013969862.1 5 - 6 857 73
XM_022419477.1 XP_022275185.1 5 - 6 787 73
GenBank mRNA AY069922.1 - 5 - 6 552 73
D29807.1 - 5 - 6 576 73

View Full Product Details