Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL2RB
Entrez Gene ID
403439
Gene Name
interleukin 2 receptor subunit beta
Gene Aliases
-
Chromosome Location
Chr.10: 27370797 - 27388775 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286850.1 NP_001273779.1 9 - 10 1140 101
NM_001286851.1 NP_001273780.1 9 - 10 1143 101
GenBank mRNA AY375181.1 - 4 - 5 398 101
DN268100.1 - 2 - 3 204 101

View Full Product Details