Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPA4
Entrez Gene ID
474680
Gene Name
heat shock protein family A (Hsp70) member 4
Gene Aliases
APG-2
Chromosome Location
Chr.11: 21290603 - 21363701 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048016.1 NP_001041481.1 16 - 17 2265 97
XM_005626399.2 XP_005626456.1 16 - 17 2100 97
XM_022424911.1 XP_022280619.1 17 - 18 2178 97
GenBank mRNA AY911512.1 - 16 - 17 2265 97

View Full Product Details