Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PODXL
Entrez Gene ID
482252
Gene Name
podocalyxin like
Gene Aliases
Gp135, PC, PCLP, PCLP-1, PCLP1, cPCLP1
Chromosome Location
Chr.14: 5614006 - 5625360 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001020809.1 NP_001018645.1 4 - 5 1145 79
GenBank mRNA AY970669.1 - 4 - 5 1145 79

View Full Product Details