Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC11C
Entrez Gene ID
404003
Gene Name
SEC11 homolog C, signal peptidase complex subunit
Gene Aliases
SEC11L3
Chromosome Location
Chr.1: 17020606 - 17036149 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003312.1 NP_001003312.1 - 732 154
GenBank mRNA J05069.1 - - 732 154

View Full Product Details