Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SEC11A
Entrez Gene ID
404004
Gene Name
SEC11 homolog A, signal peptidase complex subunit
Gene Aliases
SEC11L1, SPC18
Chromosome Location
Chr.3: 53940644 - 53980422 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003313.1 NP_001003313.1 3 - 4 341 141
XM_005618254.3 XP_005618311.1 3 - 4 423 141
GenBank mRNA J05466.1 - 3 - 4 341 141

View Full Product Details