Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HLA-DRB1
Entrez Gene ID
474860
Gene Name
MHC class II DLA DRB1 beta chain
Gene Aliases
DLA-DRB1, DLA-DRBB1, DRB, DRB1, DRBB1, LA-DRB
Chromosome Location
Chr.12: 2151409 - 2164564 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014768.1 NP_001014768.1 5 - 6 826 108
GenBank mRNA M29611.1 - 5 - 6 826 108

View Full Product Details