Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLK1
Entrez Gene ID
403942
Gene Name
kallikrein 1
Gene Aliases
-
Chromosome Location
Chr.1: 106009125 - 106028331 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003262.1 NP_001003262.1 2 - 3 210 130
GenBank mRNA X75479.1 - - 210 130

View Full Product Details