Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LMAN2
Entrez Gene ID
403938
Gene Name
lectin, mannose binding 2
Gene Aliases
VIP36
Chromosome Location
Chr.4: 36002963 - 36023788 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003258.2 NP_001003258.1 2 - 3 346 59
XM_022417332.1 XP_022273040.1 2 - 3 503 59
GenBank mRNA X76392.1 - - 347 59

View Full Product Details