Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIB2
Entrez Gene ID
403884
Gene Name
tribbles pseudokinase 2
Gene Aliases
C5FW
Chromosome Location
Chr.17: 9225598 - 9251826 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003218.1 NP_001003218.1 - 562 62
XM_005630068.3 XP_005630125.1 - 1719 62
XM_022404319.1 XP_022260027.1 - 1082 62
GenBank mRNA X99144.1 - - 562 62

View Full Product Details