Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UACA
Entrez Gene ID
403704
Gene Name
uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Aliases
C3VS
Chromosome Location
Chr.30: 34376072 - 34467278 on Build CanFam3.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003112.1 NP_001003112.1 12 - 13 1369 79
XM_005638170.3 XP_005638227.1 12 - 13 1404 79
XM_005638171.3 XP_005638228.1 12 - 13 1114 79
GenBank mRNA X99145.2 - - 1369 79

View Full Product Details